Novus Biologicals
Manufacturer Code:NBP271050UV
Catalog # NBP271050UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 134-349 of human Islet-1 (NP_002193) produced in E.coli. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Insulin gene enhancer protein ISL-1; insulin gene enhancer protein ISL-1 ISL LIM homeobox 1 Isl-1 ISL1 transcription factor LIM/homeodomain ISL1 transcription factor LIM/homeodomain (islet-1) Islet-1 ISLET1; ISL1 transcription factor, LIM/homeodomain; Islet-1
Gene Aliases: Isl-1; ISL1; ISLET1
UniProt ID: (Human) P61371
Entrez Gene ID: (Human) 3670
Molecular Function: helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.