R&D Systems
Manufacturer Code:BAF1837
Catalog # BAF1837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Islet-1 Met4-Ala349 Accession # P61371 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Insulin gene enhancer protein ISL-1; insulin gene enhancer protein ISL-1 ISL LIM homeobox 1 Isl-1 ISL1 ISL1 transcription factor LIM/homeodomain ISL1 transcription factor LIM/homeodomain (islet-1) Islet-1 ISLET1; ISL1 transcription factor, LIM/homeodomain; Islet-1
Gene Aliases: Isl-1; ISL1; ISLET1
UniProt ID: (Human) P61371
Entrez Gene ID: (Human) 3670
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.