R&D Systems
Manufacturer Code:FAB9420T100UG
Catalog # FAB9420T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 936434 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Irisin/FNDC5 Asp32-Glu143 Accession No. Q8NAU1 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: fibronectin type III domain containing 5 fibronectin type III domain-containing protein 5 FNDC5 FRCP2 FRCP2Fibronectin type III repeat-containing protein 2; Fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Irisin
Gene Aliases: FNDC5; FRCP2; irisin
UniProt ID: (Human) Q8NAU1
Entrez Gene ID: (Human) 252995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.