Novus Biologicals
Manufacturer Code:NBP259680AF488
Catalog # NBP259680AG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1] |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit PolyclonalAntibody
For Research Use Only
Protein Aliases: fibronectin type III domain containing 5 fibronectin type III domain-containing protein 5 FRCP2Fibronectin type III repeat-containing protein 2; Fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Irisin
Gene Aliases: FNDC5; FRCP2; irisin
UniProt ID: (Human) Q8NAU1
Entrez Gene ID: (Human) 252995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.