Novus Biologicals
Manufacturer Code:NB500436JF646
Catalog # NB500436646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Ds-2 |
Immunogen | Freshly ejaculated dog sperms were washed in PBS and extracted in 3% acetic acid 10% glycerol 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid lyophylized and subsequently used for immunization. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acrosomal protein SP-10; acrosomal protein SP-10 acrosomal vesicle protein 1D11S4365 SP-10 SPACA2 sperm protein 10; Acrosomal vesicle protein 1; sperm protein 10
Gene Aliases: ACRV1; D11S4365; SP-10; SPACA2
UniProt ID: (Human) P26436
Entrez Gene ID: (Human) 56
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.