Invitrogen
Manufacturer Code:MA524358
Catalog # PIMA524358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 747421 |
Immunogen | E. coli-derived recombit mouse Intelectin-2 Ala20-Ser298 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: HL-2 Intelectin-b Itln1b ITLN2 Itlnb HL-2 intelectin 2 UNQ2789/PRO7179; Intelectin-1b; Intelectin-2
Gene Aliases: Itln1b; Itln2; Itlnb
UniProt ID: (Mouse) Q80ZA0
Entrez Gene ID: (Mouse) 493583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.