R&D Systems
Manufacturer Code:FAB4775A
Catalog # FAB4775A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 416922 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Integrin beta 8 Glu43-Arg684 (predicted) Accession # P26012 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: integrin beta 8; Integrin beta-8; integrin beta-8 integrin beta 8 ITGB8; integrin, beta 8
Gene Aliases: ITGB8
UniProt ID: (Human) P26012
Entrez Gene ID: (Human) 3696
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.