Novus Biologicals
Manufacturer Code:NB2006100.025MG
Catalog # NB018132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | YFC118.3 |
Immunogen | Human neutrophils |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 959 beta subunit precursor)10 CD18 antigen CD18beta 2 cell surface adhesion glycoprotein (LFA-1 CR3 integrin beta-2 integrin beta 2 (antigen CD18 (p95) lymphocyte function-associated antigen 1macrophage antigen 1 (mac-1) beta subunit) integrin beta 2 (complement component 3 receptor 3 and 4 subunit) LAD LCAMB leukocyte cell adhesion molecule CD18 leukocyte-associated antigens CD18/11A CD18/11B CD18/11C MF17 MFI7 P150; CD18; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); Cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; Complement receptor C3 subunit beta; integrin beta chain, beta 2; Integrin beta-2; integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); leukocyte cell adhesion molecule CD18; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C
Gene Aliases: CD18; ITGB2; LAD; LCAMB; LFA-1; MAC-1; MF17; MFI7
UniProt ID: (Human) Q9UD72
Entrez Gene ID: (Human) 3689
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.