Novus Biologicals
Manufacturer Code:NBP185747
Catalog # NBP185747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:APYDDLGKVFIYHGSANGINTKPTQVLKGISPYFGYSIAGNMDLDRNSYPDVAVGSLSDSVTIFRSRPVINIQKTITVTPNRIDLRQKTACGAPSGICLQVKSCFEYTANPAGYNPSISIVGTLEAEKERRKSGLSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha6p; CD49 antigen-like family member F; CD49 antigen-like family member F CD49f CD49f antigen DKFZp686J01244 FLJ18737 integrin alpha-6 integrin alpha6B integrin alpha 6 ITGA6B VLA-6; CD49f; integrin alpha 6; Integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin, alpha 6; Processed integrin alpha-6; VLA-6
Gene Aliases: CD49f; ITGA6; ITGA6B; VLA-6
UniProt ID: (Human) P23229
Entrez Gene ID: (Human) 3655
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.