Novus Biologicals
Manufacturer Code:NBP290992
Catalog # NB025820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 5H10-27 (MFR5) |
Immunogen | C57BL/6 x A/J)F1 murine mast cell line |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD49 antigen-like family member E; CD49 antigen-like family member E CD49e CD49e antigen Fibronectin receptor subunit alpha fibronectin receptor alpha subunit FNRAVLA5A integrin alpha-5 Integrin alpha-F integrin alpha 5 (fibronectin receptor alpha polypeptide) very late activation protein 5 alpha subunit VLA-5; CD49e; Fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; integrin alpha 5; Integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); very late activation protein 5, alpha subunit; VLA-5
Gene Aliases: CD49e; FNRA; ITGA5; VLA-5; VLA5A
UniProt ID: (Human) P08648
Entrez Gene ID: (Human) 3678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.