R&D Systems
Manufacturer Code:FAB1864R100UG
Catalog # FAB1864R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 238307 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Integrin alpha 5/CD49e Phe42-Tyr995 Accession # P08648 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: CD49 antigen-like family member E; CD49 antigen-like family member E CD49e CD49e antigen Fibronectin receptor subunit alpha fibronectin receptor alpha subunit FNRAVLA5A integrin alpha-5 Integrin alpha-F integrin alpha 5 (fibronectin receptor alpha polypeptide) ITGA5 very late activation protein 5 alpha subunit VLA-5 VLA-5 alpha; CD49e; Fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; integrin alpha 5; Integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); very late activation protein 5, alpha subunit; VLA-5
Gene Aliases: CD49e; FNRA; ITGA5; VLA-5; VLA5A
UniProt ID: (Human) P08648
Entrez Gene ID: (Human) 3678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.