Novus Biologicals
Manufacturer Code:NBP197731AF405
Catalog # NBP197731W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PB36 |
Immunogen | Clone PB36 is a mouse monoclonal IgG1 kappa antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a synthetic peptide corresponding to a 32 amino acid stretch in the cytoplasmic domain of integrin alpha 3B including an appending N-terminal cysteine (CTRYYQIMPKYHAVRIREEERYPPPGSTLPTKK) coupled to keyhole limpet hemocyanin. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AA407068 CD49C GAPB3 integrin alpha 3; alpha 3 subunit of VLA-3 receptor; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; FRP-2; Galactoprotein B3; GAPB3; integrin alpha 3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); testicular tissue protein Li 96; testicular tissue protein Li 98; very late activation protein 3 receptor, alpha-3 subunit; VLA-3 subunit alpha
Gene Aliases: CD49C; FRP-2; GAP-B3; GAPB3; ILNEB; ITGA3; MSK18; VCA-2; VL3A; VLA3a
UniProt ID: (Human) P26006
Entrez Gene ID: (Human) 3675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.