R&D Systems
Manufacturer Code:FAB2787A
Catalog # FAB2787A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant mouse Integrin alpha 3/CD49c Phe33-Glu993 Accession # Q62470 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 3 subunit of VLA-3 receptor; antigen CD49C; antigen identified by monoclonal antibody J143; antigen identified by monoclonal J143 CD49 antigen-like family member C CD49c CD49c antigen FLJ34631 FLJ34704 FRP-2 Galactoprotein B3 GAP-B3 GAPB3CD49C integrin alpha-3 integrin alpha 3 (antigen CD49C alpha 3 subunit of VLA-3 receptor) ITGA3 MSK18 VCA-2 very late activation protein 3 receptor alpha-3 subunit VL3A VLA-3 subunit alpha VLA3a; CD49 antigen-like family member C; CD49c; FRP-2; Galactoprotein B3; GAPB3; integrin alpha 3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); testicular tissue protein Li 96; testicular tissue protein Li 98; very late activation protein 3 receptor, alpha-3 subunit; VLA-3 subunit alpha
Gene Aliases: CD49C; FRP-2; GAP-B3; GAPB3; ILNEB; ITGA3; MSK18; VCA-2; VL3A; VLA3a
UniProt ID: (Human) P26006
Entrez Gene ID: (Human) 3675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.