R&D Systems
Manufacturer Code:MAB12332
Catalog # MAB12332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 430903 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 subunit of VLA-2 receptor; alpha-2 subunit CD49b CD49b antigen integrin alpha 2 (CD49B alpha 2 subunit of VLA-2 receptor) ITGA2 VLA 2 VLA-2 alpha; CD49 antigen-like family member B; CD49b; Collagen receptor; GPIa; human platelet alloantigen system 5; Integrin alpha-2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); platelet antigen Br; platelet glycoprotein GPIa; Platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2 subunit alpha
Gene Aliases: BR; CD49B; GPIa; HPA-5; ITGA2; VLA-2; VLAA2
UniProt ID: (Human) P17301
Entrez Gene ID: (Human) 3673
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.