Novus Biologicals
Manufacturer Code:NBP303823100UL
Catalog # NB095648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 940-1141 of human Integrin alpha 11 (NP_001004439.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT18964 integrin alpha-11 integrin alpha 11; integrin alpha 11; Integrin alpha-11; integrin, alpha 11
Gene Aliases: HsT18964; ITGA11; MSTP018
UniProt ID: (Human) Q9UKX5
Entrez Gene ID: (Human) 22801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.