Novus Biologicals
Manufacturer Code:NBP276479
Catalog # NBP276479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL7217 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: RQISRSFFSGTQERKVQRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: CD49 antigen-like family member A; CD49 antigen-like family member A CD49a CD49a antigen integrin alpha 1 Laminin and collagen receptor very late activation protein 1 VLA-1 VLA1integrin alpha-1; CD49a; integrin alpha 1; Integrin alpha-1; integrin, alpha 1; Laminin and collagen receptor; very late activation protein 1; VLA-1
Gene Aliases: CD49a; ITGA1; VLA1
UniProt ID: (Human) P56199
Entrez Gene ID: (Human) 3672
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.