Novus Biologicals
Manufacturer Code:NBP266907
Catalog # NBP266907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JJ0949 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Abeta-degrading protease; Abeta-degrading protease EC 3.4.24 EC 3.4.24.56 FLJ35968 Insulin protease insulinase insulin-degrading enzyme Insulysin; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin
Gene Aliases: IDE; INSULYSIN
UniProt ID: (Human) P14735
Entrez Gene ID: (Human) 3416
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.