R&D Systems
Manufacturer Code:IC2496V100UG
Catalog # IC2496V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 334501 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Insulysin/IDE Met42-Leu1019 Accession # P14735 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Abeta-degrading protease; Abeta-degrading protease EC 3.4.24 EC 3.4.24.56 FLJ35968 IDE INSDEGM Insulin protease insulinase insulin-degrading enzyme Insulysin; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin
Gene Aliases: IDE; INSULYSIN
UniProt ID: (Human) P14735
Entrez Gene ID: (Human) 3416
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.