Novus Biologicals
Manufacturer Code:NBP275092
Catalog # NBP275092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence HPALPSTFRSVLGAR-C from the N Terminus of the protein sequence according to NP_004960.2 NP_001159418.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: Abeta-degrading protease; Abeta-degrading protease EC 3.4.24 EC 3.4.24.56 FLJ35968 Insulin protease insulinase insulin-degrading enzyme Insulysin; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin
Gene Aliases: IDE; INSULYSIN
UniProt ID: (Human) P14735
Entrez Gene ID: (Human) 3416
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.