Invitrogen
Catalog # PA1-29383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Neat |
Species reactivity | Human |
Host / Isotype | Guinea pig / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified human insulin conjugated to carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 10 µg/mL |
Purification | purified |
Storage buffer | proprietary buffer |
Contains | no preservative |
Storage conditions | 4° C, do not freeze |
RRID | AB_1955599 |
A recommended positive control is human pancreas.
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ins; Insulin; preproinsulin; proinsulin
Gene Aliases: IDDM; IDDM1; IDDM2; ILPR; INS; IRDN; MODY10
UniProt ID: (Human) Q5EEX2
Entrez Gene ID: (Human) 3630
Molecular Function: growth factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.