Invitrogen
Manufacturer Code:PA116988
Catalog # 01-670-141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region between residues 1-100 of human Insig1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: growth response protein (CL-6); Immediate-early protein CL-6; INSIG-1; INSIG-1 membrane protein; Insulin-induced gene 1 protein; Insulin-induced gene 1 protein INSIG-1; Insulin-induced growth response protein CL-6
Gene Aliases: CL-6; CL6; INSIG1
UniProt ID: (Human) O15503, (Rat) Q08755
Entrez Gene ID: (Human) 3638, (Rat) 64194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.