Proteintech
Manufacturer Code:175771AP150UL
Catalog # 50-173-1660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Inhibin beta B Fusion Protein Ag10956 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin AB beta polypeptide; Activin beta B chain INHBB Inhibin beta B chain inhibin beta B; Activin beta-B chain; activin betaB; Inhibin beta B chain; inhibin beta subunit B; inhibin, beta B; Inhibin, beta-2
Gene Aliases: INHBB
UniProt ID: (Human) P09529, (Mouse) Q04999, (Rat) P17491
Entrez Gene ID: (Human) 3625, (Mouse) 16324, (Rat) 25196
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.