Proteintech
Manufacturer Code:100771AP150UL
Catalog # 50-172-6004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Importin beta Fusion Protein Ag0122 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IMB1 Impnb Importin 90 Importin subunit beta 1 IPO1 IPOB karyopherin (importin) beta 1 Karyopherin subunit beta 1 KPNB1 NTF97 Nuclear factor p97 PTAC97; importin 1; importin 90; Importin beta; importin beta-1 subunit; Importin subunit beta-1; Importin-90; karyopherin (importin) beta 1; Karyopherin subunit beta-1; karyopherin,beta 1; Nuclear factor p97; Pore targeting complex 97 kDa subunit; PTAC97; SCG
Gene Aliases: AA409963; IMB1; Impnb; IPO1; IPOB; KPNB1; NTF97
UniProt ID: (Human) Q14974, (Rat) P52296, (Mouse) Q6GTI5
Entrez Gene ID: (Human) 3837, (Rat) 24917, (Mouse) 16211
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.