R&D Systems
Manufacturer Code:MAB8204SP
Catalog # MAB8204SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 843124 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Importin alpha 3/KPNA4 Met1-Asn104 Accession # O00629 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 3 Importin alpha Q1 importin subunit alpha-4 importin-alpha-Q1 IPOA3 karyopherin alpha 4 (importin alpha 3) KPNA4 MGC12217 MGC26703 Qip1 QIP12 QIP1Karyopherin subunit alpha-4 SRP3; Importin alpha Q1; Importin subunit alpha-3; importin subunit alpha-4; karyopherin alpha 4 (importin alpha 3); Karyopherin subunit alpha-4; Qip1
Gene Aliases: IPOA3; KPNA4; QIP1; SRP3
UniProt ID: (Human) O00629
Entrez Gene ID: (Human) 3840
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.