R&D Systems
Manufacturer Code:AF6207SP
Catalog # AF6207SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Importin alpha 2/KPNA2 Met1-Asp132 Accession # P52292 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: importin alpha 1 importin-alpha-P1 IPOA1 karyopherin alpha 2 (RAG cohort 1 importin alpha 1) Karyopherin subunit alpha-2 KPNA2 Pendulin QIP2 QIP2importin alpha 2 RAG cohort 1 RAG cohort protein 1 RCH1 RCH1importin subunit alpha-2 SRP1 SRP1alpha SRP1-alpha; Importin subunit alpha-1; importin subunit alpha-2; importin-alpha-P1; karyopherin alpha 2 (RAG cohort 1, importin alpha 1); Karyopherin subunit alpha-2; pendulin; RAG cohort protein 1; SRP1-alpha
Gene Aliases: IPOA1; KPNA2; QIP2; RCH1; SRP1; SRP1-alpha; SRP1alpha
UniProt ID: (Human) P52292
Entrez Gene ID: (Human) 3838
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.