Invitrogen
Manufacturer Code:PA525477
Catalog # PIPA525477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 79-106 amino acids from the N-terminal region of human IPO9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Imp9; Imp9 importin-9 ran-binding protein 9 ranBP9; imp9a; imp9b; Importin-9; importin-9a; importin-9b; Ran binding protein 9; Ran-binding protein 9; RanBP9
Gene Aliases: 0710008K06Rik; AI845704; C78347; HSPC273; IMP9; IPO9; KIAA1192; mKIAA1192; RANBP9
UniProt ID: (Human) Q96P70
Entrez Gene ID: (Human) 55705, (Mouse) 226432
Molecular Function: G-protein enzyme modulator small GTPase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.