Abnova
Manufacturer Code:H00055364B01P
Catalog # 89-017-118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | IMPACT (AAH34016 1 a.a. ~ 320 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human IMPACT protein.
For Research Use Only
Protein Aliases: Impact homolog; Imprinted and ancient gene protein homolog; MGC33718; Protein IMPACT; RWD domain containing 5
Gene Aliases: IMPACT; RWDD5
UniProt ID: (Human) Q9P2X3
Entrez Gene ID: (Human) 55364
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.