Abnova
Manufacturer Code:MAB16116
Catalog # 89-998-860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Bamboo-2 |
Immunogen | Recombinant protein corresponding to mouse mature Il1a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-1 alpha; Il-1a; Interleukin-1 alpha
Gene Aliases: Il-1a; Il1a
UniProt ID: (Mouse) P01582
Entrez Gene ID: (Mouse) 16175
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.