Proteintech
Manufacturer Code:126601AP150UL
Catalog # 50-172-8153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IkB beta Fusion Protein Ag3340 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I kappa B beta IkappaBbeta IkB B IkB beta IKBB NF kappa B inhibitor beta NF kappa BIB NFKBIB TR interacting protein 9 TRIP 9 TRIP9; I-kappa-B-beta; ikappaBbeta; IkB-B; ikB-beta; NF-kappa-B inhibitor beta; NF-kappa-BIB; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, beta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP-9
Gene Aliases: I(Kappa)B(beta); I-kappa-B-beta; IKappaBbeta; IkB; ikB-B; IKB-beta; IKBB; NF-kappa-BIB; NFKBIB; TRIP9
UniProt ID: (Human) Q15653, (Mouse) Q60778
Entrez Gene ID: (Human) 4793, (Rat) 81525, (Mouse) 18036
Molecular Function:
enzyme modulator
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.