Novus Biologicals
Manufacturer Code:NBP252518
Catalog # NBP252518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D4F2 |
Immunogen | Purified recombinant fragment of human NFKBIA (AA: 150-291) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha I-kappa-B-alpha IkappaBalphaNF-kappa-B inhibitor alpha IKBAikB-alpha IkB-alpha MAD3 MAD-3 Major histocompatibility complex enhancer-binding protein MAD3 NFKBI nuclear factor of kappa light chain gene enhancer in B-cells nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor; I-kappa-B-alpha; IkappaBalpha; IkB-alpha; Major histocompatibility complex enhancer-binding protein MAD3; NF-kappa-B inhibitor alpha; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha
Gene Aliases: IKBA; MAD-3; MAD3; NFKBI; NFKBIA
UniProt ID: (Human) P25963
Entrez Gene ID: (Human) 4792
Molecular Function: enzyme modulator hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.