Invitrogen
Manufacturer Code:MA514777
Catalog # PIMA514777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | K.79.0 |
Immunogen | Synthetic peptide corresponding to residues surrounding Asn97 of human Id2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHb26; cell growth-inhibiting gene 8; Class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID-2; DNA-binding protein inhibitor ID-2 DNA-binding protein inhibitor ID2 OTTHUMP00000140258 OTTHUMP00000200297 cell growth-inhibiting gene 8 class B basic helix-loop-helix protein 26 helix-loop-helix protein ID2 inhibitor of differentiation 2; DNA-binding protein inhibitor ID2; helix-loop-helix protein ID2; Inhibitor of differentiation 2; Inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein
Gene Aliases: AI255428; BHLHB26; C78922; GIG8; Id-2; ID2; ID2A; ID2H; Idb2
UniProt ID: (Human) Q02363, (Mouse) P41136
Entrez Gene ID: (Human) 3398, (Mouse) 15902
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.