Abnova
Manufacturer Code:MAB5609
Catalog # 89-112-316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | YN1/1.7.4 |
Immunogen | Native purified Icam1 from mouse myeloma (NS-1 cells). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rat monoclonal antibody raised against native Icam1.
For Research Use Only
Protein Aliases: CD54; ICAM-1; Intercellular adhesion molecule 1; Ly-47; MALA-2; MGC6195; MyD10
Gene Aliases: CD54; Icam-1; Icam1; Ly-47; MALA-2
UniProt ID: (Mouse) P13597
Entrez Gene ID: (Mouse) 15894
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.