Invitrogen
Manufacturer Code:PA577206
Catalog # PIPA577206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human IWS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: interacts with Spt6; IWS1 homolog; IWS1-like protein; Protein IWS1 homolog; Protein IWS1 homolog IWS1-like protein IWS1 IWS1L
Gene Aliases: 1700069O15Rik; 3000008H23; AL117927; AW214353; IWS1; IWS1L; RGD1304762
UniProt ID: (Human) Q96ST2, (Mouse) Q8C1D8, (Rat) Q3SWT4
Entrez Gene ID: (Human) 55677, (Mouse) 73473, (Rat) 291705
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.