Invitrogen
Manufacturer Code:PA5100583
Catalog # PIPA5100583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IWS1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700069O15Rik 3000008H23 AL117927 AW214353 interacts with Spt6 IWS1 IWS1 homolog IWS1 homolog (S. cerevisiae) IWS1 SUPT6H interacting protein Iws1l IWS1-like protein Protein IWS1 homolog RGD1304762; interacts with Spt6; IWS1 homolog; IWS1-like protein; Protein IWS1 homolog
Gene Aliases: 1700069O15Rik; 3000008H23; AL117927; AW214353; IWS1; IWS1L; RGD1304762
UniProt ID: (Human) Q96ST2, (Mouse) Q8C1D8, (Rat) Q3SWT4
Entrez Gene ID: (Human) 55677, (Mouse) 73473, (Rat) 291705
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.