Novus Biologicals
Manufacturer Code:NBP2851190.1ML
Catalog # NB033048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of IWS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761G0123 FLJ14655 interacts with Spt6 IWS1 homolog (S. cerevisiae) protein IWS1 homolog; interacts with Spt6; IWS1 homolog; IWS1-like protein; Protein IWS1 homolog
Gene Aliases: IWS1; IWS1L
UniProt ID: (Human) Q96ST2
Entrez Gene ID: (Human) 55677
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.