OriGene
Manufacturer Code:CF501670
Catalog # 50-166-0659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F1 |
Immunogen | Full length human recombit protein of human IVD produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACAD2; Butyryl-CoA dehydrogenase; isovaleryl CoA dehydrogenase; isovaleryl Coenzyme A dehydrogenase; Isovaleryl-CoA dehydrogenase, mitochondrial; IVD
Gene Aliases: ACAD2; IVD
UniProt ID: (Human) P26440
Entrez Gene ID: (Human) 3712
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.