Novus Biologicals
Manufacturer Code:NBP300257AF647
Catalog # NB065118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant IVD (AAH17202.1 His30-His423). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAD2 EC 1.3.99 EC 1.3.99.10 FLJ12715 FLJ34849 isovaleryl-CoA dehydrogenase mitochondrial; Butyryl-CoA dehydrogenase; isovaleryl CoA dehydrogenase; isovaleryl Coenzyme A dehydrogenase; Isovaleryl-CoA dehydrogenase, mitochondrial; IVD
Gene Aliases: ACAD2; IVD
UniProt ID: (Human) P26440
Entrez Gene ID: (Human) 3712
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.