Invitrogen
Manufacturer Code:PA579544
Catalog # PIPA579544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human ITPR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; Inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; Inositol 145-trisphosphate receptor type 3 IP3 receptor isoform 3 IP3R 3 InsP3R3 Type 3 inositol 145-trisphosphate receptor Type 3 InsP3 receptor ITPR3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R 3; IP3R-3; Type 3 inositol 1,4,5-trisphosphate receptor; Type 3 InsP3 receptor
Gene Aliases: IP3R; IP3R3; IP3R3X; Itpr-3; ITPR3
UniProt ID: (Human) Q14573, (Mouse) P70227, (Rat) Q63269
Entrez Gene ID: (Human) 3710, (Mouse) 16440, (Rat) 25679
Molecular Function:
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.