Abnova
Manufacturer Code:PAB20248
Catalog # 89-121-752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ITPR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ITPR3.
For Research Use Only
Protein Aliases: FLJ36205; inositol 1,4,5-triphosphate receptor, type 3; Inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R; IP3R 3; IP3R3; Type 3 inositol 1,4,5-trisphosphate receptor; Type 3 InsP3 receptor
Gene Aliases: IP3R; IP3R3; ITPR3
UniProt ID: (Human) Q14573
Entrez Gene ID: (Human) 3710
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.