Novus Biologicals
Manufacturer Code:NB1002466
Catalog # NB1002466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-PIEESNILSPVQD corresponding to internal region according to NP_002214.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cilia and flagella associated protein 48; Inositol 1,4,5-trisphosphate receptor type 2; inositol 145-triphosphate receptor type 2 inositol 145-trisphosphate receptor type 2 insP3R2 IP3 receptor IP3 receptor isoform 2 IP3R 2 IP3R2 Type 2 inositol 145-trisphosphate receptor Type 2 InsP3 receptor; IP3 receptor; IP3 receptor isoform 2; IP3R 2; Type 2 inositol 1,4,5-trisphosphate receptor; Type 2 InsP3 receptor
Gene Aliases: ANHD; CFAP48; INSP3R2; IP3R2; ITPR2
UniProt ID: (Human) Q14571
Entrez Gene ID: (Human) 3709
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.