Proteintech
Manufacturer Code:199621AP150UL
Catalog # 50-173-2626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; Inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate receptor, type 1; Inositol 1,4,5-trisphosphate-binding protein P400; InsP3R type I; insP3R1; INSP3R1 IP3 receptor isoform 1 IP3R IP3R 1 IP3R1 ITPR1 SCA15 SCA16 Type 1 InsP3 receptor; InsP>3<R type I; IP-3-R; IP3 receptor; IP3 receptor isoform 1; IP3R 1; opisthotonus; Protein PCD-6; protein phosphatase 1, regulatory subunit 94; Purkinje cell protein 1; Type 1 inositol 1,4,5-trisphosphate receptor; Type 1 InsP3 receptor
Gene Aliases: ACV; CLA4; D6Pas2; ENSMUSG00000072853; Gm10429; I145TR; InsP3R; INSP3R1; IP3R; IP3R1; Itpr-1; ITPR1; opt; P400; Pcd6; Pcp-1; Pcp1; PPP1R94; SCA15; SCA16; SCA29
UniProt ID: (Human) Q14643, (Mouse) P11881, (Rat) P29994
Entrez Gene ID: (Human) 3708, (Mouse) 16438, (Rat) 25262
Molecular Function: ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.