Proteintech
Manufacturer Code:128161AP150UL
Catalog # 50-172-8271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ITPKB Fusion Protein Ag3456 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol 1,4,5-trisphosphate 3-kinase B; Inositol-trisphosphate 3-kinase B; InsP 3 kinase B IP3 3 kinase B IP3K IP3K B IP3KB ITPKB PIG37; insP 3-kinase B; IP3 3-kinase B; IP3K B; IP3K-B; proliferation-inducing protein 37
Gene Aliases: 1110033J02Rik; E130307H12Rik; IP3-3KB; IP3K; IP3K-B; IP3KB; ITPKB; PIG37
UniProt ID: (Human) P27987, (Rat) P42335
Entrez Gene ID: (Human) 3707, (Mouse) 320404, (Rat) 54260
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.