Invitrogen
Manufacturer Code:PA585786
Catalog # PIPA585786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse IP3KA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: inositol 1,4,5-triphosphate 3-kinase; Inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; Inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3-kinase A; IP3-kinase A; IP3K A; Itpka antibody inositol 145-trisphosphate 3-kinase A antibody
Gene Aliases: Itpka
UniProt ID: (Mouse) Q8R071, (Rat) P17105
Entrez Gene ID: (Mouse) 228550, (Rat) 81677
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.