Invitrogen
Manufacturer Code:PA5101093
Catalog # PIPA5101093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IP3KA corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.127 inositol 145-triphosphate 3-kinase inositol 145-trisphosphate 3-kinase A inositol 145-triphosphate 3-kinase inositol-trisphosphate 3-kinase A insP 3-kinase A IP3 3 kinase A IP3 3-kinase A IP3-3KA IP3K A IP3KA IP3-kinase A ITPKA; inositol 1,4,5-triphosphate 3-kinase; Inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; Inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3-kinase A; IP3-kinase A; IP3K A
Gene Aliases: IP3-3KA; IP3KA; ITPKA
UniProt ID: (Human) P23677, (Mouse) Q8R071, (Rat) P17105
Entrez Gene ID: (Human) 3706, (Mouse) 228550, (Rat) 81677
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.