Novus Biologicals
Manufacturer Code:NBP304719100UL
Catalog # NB095089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 141-461 of human ITPKA (NP_002211.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.127 inositol 145-trisphosphate 3-kinase AIP3 3-kinase A inositol-trisphosphate 3-kinase A InsP 3-kinase A IP3-3KA IP3K A IP3KA; Inositol 1,4,5-trisphosphate 3-kinase A; Inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3-kinase A; IP3K A
Gene Aliases: IP3-3KA; IP3KA; ITPKA
UniProt ID: (Human) P23677
Entrez Gene ID: (Human) 3706
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.