Novus Biologicals
Manufacturer Code:NBP245463
Catalog # NBP245463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F5 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 309-391 of human ITPK1(NP_001136065) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.134 EC 2.7.1.159 inositol 134-triphosphate 5/6 kinase inositol 134-trisphosphate 5/6 kinase Inositol 134-trisphosphate 5/6-kinase inositol-tetrakisphosphate 1-kinase Inositol-triphosphate 5/6-kinase Ins(134)P(3) 5/6-kinase ITRPK1; Inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; Inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase
Gene Aliases: ITPK1; ITRPK1
UniProt ID: (Human) Q13572
Entrez Gene ID: (Human) 3705
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.