Invitrogen
Manufacturer Code:PA521883
Catalog # PIPA521883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 171 and 414 of ITPK1 (Uniprot IDNo.Q13572) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; Inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase; ITRPK1 inositol 134-triphosphate 5/6 kinase inositol 134-trisphosphate 5/6 kinase inositol 134-trisphosphate 5/6-kinase inositol-triphosphate 5/6-kinase ins(134)P(3) 5/6-kinase
Gene Aliases: BC031182; ITPK1; ITRPK1
UniProt ID: (Human) Q13572, (Mouse) Q8BYN3
Entrez Gene ID: (Human) 3705, (Mouse) 217837
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.