Novus Biologicals
Manufacturer Code:NBP132494
Catalog # NBP132494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ITPase. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf37 dJ794I6.3 EC 3.6.1 EC 3.6.1.19 HLC14-06-P Inosine triphosphatase inosine triphosphatase (nucleoside triphosphate pyrophosphatase) inosine triphosphatase-A inosine triphosphate pyrophosphatase inosine triphosphate pyrophosphohydrolase ITPase My049 protein nucleoside triphosphate diphosphatase Putative oncogene protein hlc14-06-p; inosine triphosphatase (nucleoside triphosphate pyrophosphatase); Inosine triphosphate pyrophosphatase; inosine triphosphate pyrophosphohydrolase; ITPase; Non-canonical purine NTP pyrophosphatase; Non-standard purine NTP pyrophosphatase; NTPase; Nucleoside-triphosphate diphosphatase; Nucleoside-triphosphate pyrophosphatase; Putative oncogene protein hlc14-06-p
Gene Aliases: C20orf37; dJ794I6.3; HLC14-06-P; ITPA; ITPase; My049; NTPase; OK/SW-cl.9
UniProt ID: (Human) Q9BY32
Entrez Gene ID: (Human) 3704
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.