Invitrogen
Manufacturer Code:PA564102
Catalog # PIPA564102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ITLN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endothelial lectin HL-1; Galactofuranose-binding lectin; HL-1 HL1 INTL ITLN LFR hIntL omentin ITLN-1 endothelial lectin HL-1 galactofuranose-binding lectin intelectin-1 intestinal lactoferrin receptor; intelectin 1 (galactofuranose binding); Intelectin-1; Intestinal lactoferrin receptor; ITLN-1; Omentin
Gene Aliases: hIntL; HL-1; HL1; INTL; ITLN; ITLN1; LFR; omentin; UNQ640/PRO1270
UniProt ID: (Human) Q8WWA0
Entrez Gene ID: (Human) 55600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.