Abnova
Manufacturer Code:RAB00564
Catalog # 89-339-520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ITGB3/3126R |
Immunogen | Recombinant protein corresponding to amino acids 385-490 of human ITGB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C |
Rabbit Monoclonal Antibody.
Protein Aliases: antigen CD61; CD61; CD61 GP3A GPIIIa; GPIIIa; integrin beta 3; Integrin beta-3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa
Gene Aliases: BDPLT16; BDPLT2; CD61; GP3A; GPIIIa; GT; ITGB3
UniProt ID: (Human) P05106
Entrez Gene ID: (Human) 3690
Molecular Function:
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.